63401-001
STRboost®
50 rxn, for multiplexing PCR samples
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
form
dry
usage
50 reactions
Storage temp.
room temp
General description
STRboost® is a novel reagent specifically formulated to enhance multiplex PCR amplification reactions used for human identification applications. STRboost® can overcome limitations associated with multiplex STR analysis performed on compromised sample types. Use of STRboost® resolves stochastic effects such as peak balance issues, allele dropout and sub-optimal peak height definition.
Don′t see what you are looking for? Custom stabilization services for molecular diagnostic assay and reagent manufacturers are available through Sigma′s partner, Biomatrica. PCRstable® enables the preservation of PCR-based assay components in any format (liquid or dry).
Don′t see what you are looking for? Custom stabilization services for molecular diagnostic assay and reagent manufacturers are available through Sigma′s partner, Biomatrica. PCRstable® enables the preservation of PCR-based assay components in any format (liquid or dry).
Packaging
1 bottle
Physical form
50 reactions (150 μL) in screw-top microfuge tube made of durable polypropylene.
Legal Information
PCRstable is a registered trademark of Biomatrica
STRboost is a registered trademark of Biomatrica
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service