Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

62660

Sigma-Aldrich

LR Accelerator

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

impurities

≥20% N,N-dimethyl-p-toluidine

density

1.050 g/cm3

application(s)

hematology
histology

storage temp.

room temp

Application

LR Accelerator is used for the chemical curing of LR White and LR Gold embedding media for routine and low temperature electron microscopy or the immunocytochemical localization of antigens by either light or electron microscopy.

related product

Product No.
Description
Pricing

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - Repr. 2 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Reproductive organs

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

168.8 °F

flash_point_c

76 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margarita Sobol et al.
Histochemistry and cell biology, 135(1), 103-110 (2010-12-15)
The best available approach of biological sample preparation for transmission electron microscopy currently includes cryoimmobilization by high-pressure freezing (HPF) followed by freeze-substitution (FS). This method has been well established for interphase cells; however, a reliable and easy procedure is still
Olivier Gros et al.
Acta histochemica, 110(5), 427-431 (2008-01-12)
Hydrophilic resins present the advantage of making possible both hybridization experiments involving either antibodies or oligonucleotide probes and ultrastructural observations. Whereas various embedding protocols are available, only very few concern flat-embedded preparations. In this study we describe an easy protocol
Vendula Strádalová et al.
Histochemistry and cell biology, 130(5), 1047-1052 (2008-09-18)
A protocol for high-pressure freezing and LR White embedding of mammalian cells suitable for fine ultrastructural studies in combination with immunogold labelling is presented. HeLa S3 cells enclosed in low-temperature gelling agarose were high-pressure frozen, freeze-substituted in acetone, and embedded
Shuji Yamashita
Methods in molecular biology (Clifton, N.J.), 657, 237-248 (2010-07-06)
We describe a standardized method of fixation, antigen retrieval, and image contrasting for post-embedding immunoelectron microscopy. Tissues are fixed with formaldehyde solutions containing Ca(2+) and Mg(2+) ions at pH 7.4 and then at pH 8.5. After dehydration with dimethylformamide, the
Margarita Sobol et al.
Histochemistry and cell biology, 134(6), 631-641 (2010-11-11)
In this study we present an optimized method of high-pressure freezing and automated freeze-substitution of cultured human cells, followed by LR White embedding, for subsequent immunolabeling. Also, the influence of various conditions of the freeze-substitution procedures such as temperature, duration

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service