Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

62579

Sigma-Aldrich

Lithium perchlorate

BioUltra, ≥99.0% (calc. based on dry substance, T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiClO4
CAS Number:
Molecular Weight:
106.39
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioUltra

assay

≥99.0% (calc. based on dry substance, T)

impurities

insoluble matter, passes filter test
≤3% water

pH

6.0-9.5 (25 °C, 1 M in H2O)

mp

236 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤20 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.037
λ: 280 nm Amax: ≤0.032

SMILES string

[Li+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Li/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

MHCFAGZWMAWTNR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lithium perchlorate may be useful as a component in the development of biopolymers and biofilms. Lithium perchlorate is a useful reagent in synthetic processes and protein analysis.

Physical form

powder or fine crystals

Other Notes

An effective protein denaturant

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Fuentes et al.
Biomacromolecules, 1(2), 239-243 (2001-11-17)
Chitosan/poly(aminopropylsiloxane) hybrid films were obtained by blending 3-(aminopropyl)siloxane oligomers (pAPS) with chitosan (CHI). The pAPS oligomers were prepared by the sol-gel method starting from 3-(aminopropyl)triethoxysilane. These hybrids were characterized by chemical, spectroscopic and morphological methods. Scanning electron micrographs of hybrid
S Maruyama et al.
Biochimica et biophysica acta, 494(2), 343-353 (1977-10-26)
In an attempt to understand the specific effect of inorganic protein denaturants, lithium cation and perchlorate anion, upon the molecular conformation of bovine alpha-lactalbumin and to characterize the denatured states of the protein and the denaturation processes, themodynamic studies on
A comparison of the denatured states of alpha-lactalbumin and lysozyme.
R N Sharma et al.
Journal of molecular biology, 88(1), 247-257 (1974-09-05)
J Torres et al.
Biophysical journal, 68(5), 2049-2055 (1995-05-01)
Fourier transform infrared and UV fourth-derivative spectroscopies were used to study the secondary structure of bacteriorhodopsin and its chymotryptic and one of the sodium borohydride fragments dissolved in chloroform-methanol (1:1, v/v), 0.1 M LiClO4. The C1 fragment (helices C, D
S Selvasekarapandian et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 65(5), 1234-1240 (2006-07-11)
The polymer electrolytes composed of poly(vinyl acetate) (PVAc) with various stoichiometric ratios of lithium perchlorate (LiClO(4)) salt have been prepared by solution casting method. The techniques Fourier transform infra-red (FTIR) and Laser Raman spectroscopy have been used to monitor polymer-salt

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service