Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

62497

Sigma-Aldrich

Lithium fluoride

BioUltra, ≥99.0% (F)

Synonym(s):

Fluorolithium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiF
CAS Number:
Molecular Weight:
25.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

assay

≥99.0% (F)

form

powder

impurities

insoluble matter, passes filter test

pH

6.0-8.5 (25 °C, 0.01 M in H2O)

mp

845 °C (lit.)

solubility

H2O: 0.01 M at 20 °C, clear, colorless

density

2.64 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Al: ≤200 mg/kg
As: ≤5 mg/kg
Ba: ≤50 mg/kg
Bi: ≤20 mg/kg
Ca: ≤500 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤10 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Mg: ≤100 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤100 mg/kg
Pb: ≤10 mg/kg
Sr: ≤100 mg/kg
Zn: ≤10 mg/kg

λ

0.01 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.01
λ: 280 nm Amax: ≤0.01

SMILES string

[Li+].[F-]

InChI

1S/FH.Li/h1H;/q;+1/p-1

InChI key

PQXKHYXIUOZZFA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lithium fluoride, the lithium salt of hydrofluoric acid, may be used as an adjuvant in antibiotic therapy. Lithium fluoride is useful for the development of nanostructures and nanolayers.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xing Wu et al.
ACS nano, 14(7), 9125-9135 (2020-06-27)
The design of nanosheets interlayer between the substrate and polyamide layer has attracted growing attention to improve the performance of thin-film composite membranes. However, the membrane size is limited by current fabrication methods such as vacuum filtration. Herein, a high-performance
X W Meng et al.
The journal of physical chemistry. B, 115(16), 4768-4773 (2011-04-06)
On the basis of molecular dynamics simulations, we investigate water permeation across a single-walled carbon nanotube (SWCNT) under the influence of four symmetrical half-rings, each having six LiF dipolar molecules. The flux remains almost fixed as the separation, R, between
Christian Bernhardsson et al.
Health physics, 103(6), 740-749 (2012-11-01)
Laboratory studies have shown that ordinary household salt (NaCl) exhibits several promising properties for retrospective dosimetry; e.g., a linear dose response and a low detection limit, down to a few mGy or even less. In an attempt to test NaCl
Hatem Dachraoui et al.
Optics express, 19(3), 2797-2804 (2011-03-04)
We report a femtosecond crystallographic study of the dependence of the free-carries generation to the alignment of a crystalline sample to the laser polarization. The probe pulse transmission exhibits a π/2 modulation that is shown to be correlated with the
Thorsten Grünheid et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 141(4), 436-443 (2012-04-03)
Cone-beam computed tomography (CBCT) has become a routine imaging modality for many orthodontic clinics. However, questions remain about the amount of radiation patients are exposed to during the scans. This study determined the amounts of radiation potentially absorbed by a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service