Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61843

Sigma-Aldrich

α-1,3-Fucosyltransferase kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

shipped in

dry ice

storage temp.

−20°C

Application

This kit contains sufficient amounts of enzyme, nucleotide sugar (donor), buffer and supplementary reagents for performing five glycosylation reactions.

Kit Components Only

Product No.
Description

  • α-1,3-Fucosyl Transferase VI recombinant from Pichia pastoris, ≥1.0 U/mL

  • Manganese(II) chloride tetrahydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)

  • Phosphatase, Alkaline from bovine intestinal mucosa, solution (clear), >10000 U/mL

Kit Components Also Available Separately

Product No.
Description
SDS

  • 05470Bovine Serum Albumin, lyophilized powder, crystallized, ≥98.0% (GE)SDS


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service