Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61707

Sigma-Aldrich

Streptavidin C-Phycocyanin

Synonym(s):

Phycocyanin-Streptavidin C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

fluorescence

λex 600 nm; λem 640 nm in 0.1 M phosphate pH 7.2

Storage temp.

−20°C

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.
One vial contains 0.5 mg of conjugate and ~11 mg of additives.

Other Notes

Molar ratio of pigment:streptavidin is (1:2.5).

Reconstitution

Reconstitute one vial to 250 μl with distilled or deionized water (the solution will consist of 100 mM sodium phosphate pH 7.2, 150 mM sodium chloride, 50 mM sucrose, 0.1% BSA, 0.05% sodium azide and 2 mg/ml pigment). Do not freeze, store at 4 °C in the dark.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service