Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59505

Sigma-Aldrich

β-1,4-Galactosyl Transferase Kit

Synonym(s):

β-1,4-Galactosylation Kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352204

storage temp.

−20°C

Components

This kit contains sufficient amounts of glycosyltransferase, nucleotide sugar (donor), buffer and supplementary reagents for performing five glycosylations.

Kit Components Only

Product No.
Description

  • β-1,4-Galactosyl Transferase from bovine milk, ≥0.6 U/mg

  • Manganese(II) chloride tetrahydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)

  • α-Lactalbumin from bovine milk, calcium depleted, ≥90% (GE)

  • Phosphatase, Alkaline from bovine intestinal mucosa, solution (clear), >10000 U/mL

Kit Components Also Available Separately

Product No.
Description
SDS

  • Trizma® Buffer substance, pH 7.4

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

target_organs

Brain

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Gebhardt et al.
Helvetica Chimica Acta, 85, 1943-1943 (2002)
B Danieli et al.
The Journal of organic chemistry, 66(1), 262-269 (2001-06-30)
Ginsenoside Rg1 (1), the most representative Ginsenoside from Panax Ginseng C.A. Meyer belonging to the protopanaxatriol family, has been galactosylated by action of the beta-(1,4)-galactosyltransferase (GalT) from bovine colostrum, using UDP-galactose as an activated sugar donor. The enzyme showed the
S Riva et al.
Carbohydrate research, 305(3-4), 525-531 (1998-07-02)
The influence of various organic cosolvents on the stability and activity of the beta-1,4-galactosyltransferase from bovine colostrum (GalT) and of its ancillary enzyme UDP-galactose-4'-epimerase has been investigated using the glucosylated alkaloid colchicoside (1) as a model substrate. It has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service