Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59427C

SAFC

Trypsin Solution 10X

Synonym(s):

Trypsin solution from porcine pancreas

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

sterility

sterile; sterile-filtered

form

liquid

quality

Trypsin Solution 10X is a 2.5% solution of 1:250 trypsin

components

phenol red: 10.62 mg/L

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The 1:250 designation indicates the tryptic activity based on standardized testing. This ratio indicates that one part trypsin will digest 250 parts casein in the USP activity assay.
Trypsin from porcine pancreas is an enzyme that is used for the dissociation and dislodging of mammalian cells and tissues that are anchorage-dependent. The age of the culture and cell type determines the concentration of trypsin necessary to disaggregate cells from their substrate.

application

Trypsin Solution 10X has been used to release adherent cells from tissue culture plates for passaging.[1]

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xufang Deng et al.
Journal of virology, 94(17) (2020-06-20)
Coronaviruses (CoVs) have repeatedly emerged from wildlife hosts and infected humans and livestock animals to cause epidemics with significant morbidity and mortality. CoVs infect various organs, including respiratory and enteric systems, as exemplified by newly emerged severe acute respiratory syndrome
Utilizing Combined Methodologies to Define the Role of Plasma Membrane Delivery During Axon Branching and Neuronal Morphogenesis
Journal of Visualized Experiments, 109 (2016)
Aniella Abi-Gerges et al.
Pulmonary pharmacology & therapeutics, 63, 101940-101940 (2020-09-06)
Waterpipe smoking is a popular pastime worldwide with statistics pointing to an alarming increase in consumption. In the current paper, the evaluation of sub-chronic waterpipe smoke exposure was undertaken using C57BL/6 female mice using a dynamic exposure setting to emulate
Wenfeng Liu et al.
Experimental physiology, 104(5), 715-728 (2019-03-02)
What is the central question of this study? What is the biological role of carbonylation in muscle age-related functional decline and how might exercise affect the carbonylation process differently compared to habitual sedentary behaviour? What is the main finding and
Carolina P Bernardes et al.
Peptides, 104, 24-34 (2018-04-24)
Venom small peptides that target neurotrophin receptors might be beneficial in neurodegeneration, including Parkinsońs disease (PD). Their small size, ease of synthesis, structural stability and target selectivity make them important tools to overcome the limitations of endogenous neurotrophins as therapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service