Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56733

Sigma-Aldrich

IEF-Marker 3.6-9.3

mixture of 8 proteins, lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105336

form

solid

quality

lyophilized

suitability

in accordance for electrophoresis test

storage temp.

2-8°C

Packaging

1 piece contains ~0.9 mg marker protein and glycine

Components

Composition (marker protein, pI):
1. amyloglucosidase from aspergillus niger, 3.6
2. trypsin inhibitor from soybean, 4.6
3. β-lactoglobulin A from bovine milk, 5.1
4. carbonic anhydrase II from bovine erythrocytes, 5.9
5. carbonic anhydrase I from human erythrocytes, 6.6
6. myoglobin from heart, 6.8, 7.2
7. lentil lecitin from lens culinaris, 8.2, 8.6, 8.8
8. trypsinogen from bovine pancreas, 9.3

Reconstitution

After reconstitution with 250 μl H2O 1 piece contains 0.2 M glycine pH ~6. For gel electrophoresis please use 10 μl of marker mixture per lane.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service