Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56612

Sigma-Aldrich

InP/ZnS nanoparticles

core-shell type, green emitting, ~1 mg/mL

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352202

concentration

~1 mg/mL

fluorescence

λem 530 nm±10 nm

Looking for similar products? Visit Product Comparison Guide

Application

InP/ZnS nanoparticles, core-shell type, green emitting is part of a family of InP/ZnS nanoparticles intended for use in molecular imaging applications and quantum dot development.

Analysis Note

full width at half maximum ≤100 nm

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F - closed cup - Solvent

flash_point_c

4.0 °C - closed cup - Solvent

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Gao et al.
Optics express, 19(6), 5528-5535 (2011-03-30)
High-quality InP/ZnS core-shell nanocrystal quantum dots (NQDs) were synthesized as a heavy-metal-free alternative to the gain media of cadmium-based colloidal nanoparticles. Upon UV excitation, amplified spontaneous emission (ASE) and optical gain were observed, for the first time, in close-packed InP/ZnS
Reverse type-I ZnSe/InP/ZnS core/shell/shell nanocrystals: cadmium-free quantum dots for visible luminescence.
Sungwoo Kim et al.
Small (Weinheim an der Bergstrasse, Germany), 7(1), 70-73 (2010-12-07)
Jinhao Gao et al.
Nano letters, 12(1), 281-286 (2011-12-17)
The use of quantum dots (QDs) in biomedical research has grown tremendously, yet successful examples of clinical applications are absent due to many clinical concerns. Here, we report on a new type of stable and biocompatible dendron-coated InP/ZnS core/shell QD
Sudarsan Tamang et al.
ACS nano, 5(12), 9392-9402 (2011-11-01)
Small thiol-containing amino acids such as cysteine are appealing surface ligands for transferring semiconductor quantum dots (QDs) from organic solvents to the aqueous phase. They provide a compact hydrodynamic diameter and low nonspecific binding in biological environment. However, cysteine-capped QDs
Full visible range covering InP/ZnS nanocrystals with high photometric performance and their application to white quantum dot light-emitting diodes.
Xuyong Yang et al.
Advanced materials (Deerfield Beach, Fla.), 24(30), 4180-4185 (2012-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service