Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56177

Sigma-Aldrich

Hyaluronate Lyase from Streptococcus pyogenes

≥8.0 units/mg protein (5.0-15.0 mg/mL)

Synonym(s):

Hyaluronidase from Streptococcus pyogenes

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

form

liquid

specific activity

≥8.0 units/mg protein (5.0-15.0 mg/mL)

storage temp.

2-8°C

Biochem/physiol Actions

Hyaluronate lyase degrades hyaluronate using an elimination reaction to break glycosidic linkages yielding unsaturated disaccharide products. It is important for the diffusion of toxins and proteins produced by S. pyogenes.
Hyaluronate lyase degrades hyaluronate using an elimination reaction to break glycosidic linkages yielding unsaturated disaccharide products.[1] It is important for the diffusion of toxins and proteins produced by S. pyogenes.[2]

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol unsaturated disaccharides per minute at pH 6.0 and 37°C from hyaluronic acid (Product No. 53747).

Physical form

ammonium sulfate suspension

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural studies on the bacterial lyase-resistant tetrasaccharides derived from the antithrombin III-binding site of porcine intestinal heparin.
Yamada, S., et al.
The Journal of Biological Chemistry, 4780-4787 (1993)
Renata K T Kobayashi et al.
Journal of microbiological methods, 68(3), 654-655 (2006-12-13)
The temperature-sensitive hemagglutinin (Tsh, 140 kDa) produced by Escherichia coli is cleaved into a fragment (106 kDa) containing mucinase activity, and an agglutinin fragment (33 kDa). By incorporating mucins into SDS-PAGE gels stained by Schiff's periodic acid, we could simultaneously
Parul Mishra et al.
The FEBS journal, 276(12), 3392-3402 (2009-05-15)
Hyaluronate lyases are a class of endoglycosaminidase enzymes with a high level of complexity and heterogeneity. The main function of the Streptococcus pyogenes bacteriophage protein hyaluronate lyase, HylP2, is to degrade hyaluronan into unsaturated disaccharide units. HylP2 was cloned, over-expressed
Clarise Rivera Starr et al.
Infection and immunity, 74(1), 40-48 (2005-12-22)
Group A streptococcus (GAS) depends on a hyaluronic acid (HA) capsule to evade phagocytosis and to interact with epithelial cells. Paradoxically, GAS also produces hyaluronidase (Hyl), an enzyme that cleaves HA. A common assumption is that Hyl digests structurally identical
Carlos Martinez-Fleites et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 65(Pt 10), 963-966 (2009-10-24)
The crystal structures of truncated forms of the Streptococcus pyogenes phage-encoded hyaluronate lyases HylP2 and HylP3 were determined by molecular replacement to 1.6 and 1.9 A resolution, respectively. The truncated forms crystallized in a hexagonal space group, forming a trimer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service