Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55548

Sigma-Aldrich

Adenosine 2′,3′-cyclic phosphate 5′-phosphosulfate triethylammonium salt

≥85% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O12P2S · xC6H15N
CAS Number:
Molecular Weight:
489.25 (free acid basis)
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

assay

≥85% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CCN(CC)CC.Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OS(O)(=O)=O)[C@H]4OP(O)(=O)O[C@@H]34

InChI

1S/C10H13N5O12P2S.C6H15N/c11-8-5-9(13-2-12-8)15(3-14-5)10-7-6(25-29(18,19)26-7)4(24-10)1-23-28(16,17)27-30(20,21)22;1-4-7(5-2)6-3/h2-4,6-7,10H,1H2,(H,16,17)(H,18,19)(H2,11,12,13)(H,20,21,22);4-6H2,1-3H3/t4-,6-,7-,10-;/m1./s1

InChI key

QSKHVLWLKHDUCX-MCDZGGTQSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service