Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55536

Sigma-Aldrich

Fluorescent Red Mega 480 NHS-ester

suitable for fluorescence, ≥90% (coupling to amines)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H33N3O9S
Molecular Weight:
611.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

assay

≥90% (coupling to amines)

fluorescence

λex 513 nm; λem 640 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CCN(CC)c1ccc2C=C(\C=C\c3ccc(c[n+]3CCCCCC(=O)ON4C(=O)CCC4=O)S([O-])(=O)=O)C(=O)Oc2c1

InChI

1S/C30H33N3O9S/c1-3-31(4-2)24-12-9-21-18-22(30(37)41-26(21)19-24)10-11-23-13-14-25(43(38,39)40)20-32(23)17-7-5-6-8-29(36)42-33-27(34)15-16-28(33)35/h9-14,18-20H,3-8,15-17H2,1-2H3

InChI key

JLMODFISYHYYEO-UHFFFAOYSA-N

Application

Fluorescent Red Mega 480 NHS-ester is an amine conjugating fluorescent biolabel that can be used for covalent coupling to proteins and other biomolecules containing primary amino groups. Fluorescent red Mega 480 NHS ester and other Mega labels are specifically designed for multicolor techniques. It is characterized by an extremely large stoke′s shift between excitation and emission maxima. This label is especially well suited for excitation by argon lasers or other short wavelength excitation light. This means that it can be excited with the same light source or filter arrangement than fluorescein and other widely used dyes as well as other Mega labels. Due to the different emission wavelength several different labels can be detected independently in the same experiment. Due to the small size of the Mega-labels and the direct conversion of excitation light into emission without using a FRET mechanism we foresee a variety of possible applications including DNA sequencing and FISH microscopy.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service