Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55183

Sigma-Aldrich

NTA -Tracy 652

suitable for fluorescence, ≥80% (HPCE)

Synonym(s):

Nα, Nα-bis(carboxymethyl)-L-lysine Nickel(II) complex - Tracy 652 conjugate, Tracy 652 - Nitrilotriacetic acid, complexed to Ni2+ - ion

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

assay

≥80% (HPCE)

fluorescence

λex 662 nm; λem 683 nm in PBS

suitability

suitable for fluorescence

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Tracy 652 is a newly developed NIR fluorescent Dye. The spectral range above 650 nm is of particular interested in biology analysis and imaging due to the reduced background and improved sensitivity.
Tracy 652 is highly water-soluble and stable dye.

Application

for the fluorescence detection of HIS-tagged proteins

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service