Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55171

Sigma-Aldrich

(23R)-23,25-Dihydroxyvitamin D3

≥99.0% (HPLC)

Synonym(s):

(23R)-23,25-Dihydroxycholecalciferol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

assay

≥99.0% (HPLC)

form

powder or crystals

color

colorless to white

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@H](C[C@@H](O)CC(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C3\C[C@@H](O)CCC3=C

InChI

1S/C27H44O3/c1-18-8-11-22(28)16-21(18)10-9-20-7-6-14-27(5)24(12-13-25(20)27)19(2)15-23(29)17-26(3,4)30/h9-10,19,22-25,28-30H,1,6-8,11-17H2,2-5H3/b20-9+,21-10-/t19-,22+,23-,24-,25+,27-/m1/s1

InChI key

JVBPQHSRTHJMLM-WORHRCAZSA-N

Application

able to increase the plasma lactone concentration[1]

Biochem/physiol Actions

Cholecalciferol is an inactive form of vitamin D3 which undergoes various levels of hydroxylation to form active vitamin D3 analogs. 1α-Hydroxyvitamin D3 (alfacalcidol) is a synthetic analog that is metabolized to 1,25-dihydroxycholecalciferol, the biologically active form of vitamin D3. Other analogues of cholecalciferol result from different hydroxylations. 23,25-Dihydroxyvitamin D3 is synthesized by the human placenta and by kidney cortex mitochondria.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Shany et al.
Nephron, 42(2), 141-145 (1986-01-01)
We measured 24,25-dihydroxyvitamin D [24,25(OH)2D] levels in patients treated with chronic ambulatory peritoneal dialysis (CAPD), before and after receiving vitamin D2 or 1 alpha-hydroxyvitamin D3 (1 alpha-OH-D3). Vitamin D2 administration led to an increase in 25-hydroxyvitamin D (25-OH-D) and a
R L Horst et al.
Biochemistry, 22(2), 245-250 (1983-01-18)
A new metabolite of 23,25-dihydroxyvitamin D3 has been generated with kidney homogenates prepared from vitamin D treated chicks. The metabolite was purified with three high-performance liquid chromatographic steps and was identified as 23-keto-25-hydroxyvitamin D3 by ultraviolet absorption spectroscopy, mass spectrometry
S Ishizuka et al.
Archives of biochemistry and biophysics, 228(1), 179-184 (1984-01-01)
To elucidate the biosynthesis of 25-hydroxyvitamin D3-26,23-lactone, various vitamin D3 derivatives were incubated individually with kidney homogenates prepared from vitamin D3-supplemented chicks, a preparation known to produce the 25-hydroxyvitamin D3-26,23-lactone from 25-hydroxyvitamin D3. The 25-hydroxyvitamin D3-26, 23-lactone produced in vitro
G Jones et al.
Biochemistry, 23(16), 3749-3754 (1984-07-31)
By cochromatography, mass spectrometry, and chemical derivatization, we have shown that a metabolite isolated from the perfused rat kidney incubated with 24-(R),25-dihydroxyvitamin D3 is indistinguishable from chemically synthesized 24,25,26,27-tetranor-23-hydroxyvitamin D3. The new metabolite is also produced from 24-oxo-25-hydroxyvitamin D3 but
J Z Zhang et al.
The Journal of dermatology, 22(5), 305-309 (1995-05-01)
Platelet-derived growth factor (PDGF) is a potent mitogen for several mesenchymal cells and plays an important role in wound repair. Three PDGF isoforms, PDGF-AA, PDGF-BB, and PDGF-AB, have been found to be generated in various tissues. PDGF-AB production by normal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service