Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54367

Sigma-Aldrich

Phalloidin-Atto 520

suitable for fluorescence

Synonym(s):

Atto 520-Phalloidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

form

solid

mol wt

Mw 1250 g/mol

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 516 nm; λem 538.0 nm±10.0 nm in PBS, pH 7.4

suitability

suitable for fluorescence

Storage temp.

−20°C

General description

Atto 520 is a novel fluorescent label related to the well-known dye Rhodamine 6G. Characteristic features of the label are strong absorption, high fluorescence quantum yield, and high thermal and photo-stability. The dye is moderately hydrophilic. Above pH 7.0 the dye exists in an equilibrium with a colourless form (pseudobase). This is a reversible reaction, which does not interfere with the process of coupling to proteins etc. On lowering the pH absorption and fluorescence are restored fully. For details of coupling see our recommended labeling procedures The fluorescence is excited most efficiently in the range 510 - 535 nm. A suitable source of excitation is an Argon-Ion laser using the 514 nm line.
Phalloidin is a fungal toxin isolated from the poisonous mushroom Amanita phalloides. Its toxicity is attributed to the ability to bind F actin in liver and muscle cells. As a result of binding phalloidin, actin filaments become strongly stabilized. Phalloidin has been found to bind only to polymeric and oligomeric forms of actin, and not to monomeric actin. The dissociation constant of the actin-phalloidin complex has been determined to be on the order of 3 x 10-8. Phalloidin differs from amanitin in rapidity of action; at high dose levels, death of mice or rats occurs within 1 or
2 hours. Fluorescent conjugates of phalloidin are used to label actin filaments for histological applications. Some structural features of phalloidin are required for the binding to actin. However, the side chain of amino acid 7 (g-d-dihydroxyleucine) is accessible for chemical modifications without appreciable loss of affinity for actin.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service