53651
1-[3-(Succinimidyloxycarbonyl)benzyl]-4-[2-(3,4-dihydro-2H-1-benzopyran-6-yl)-5-oxazolyl]pyridinium bromide
BioReagent, suitable for fluorescence, ≥90% (coupling to amines)
About This Item
product line
BioReagent
assay
≥90% (coupling to amines)
form
solid
fluorescence
λex 387 nm; λem 536 nm in 0.1 M phosphate pH 7.0
suitability
suitable for fluorescence
Storage temp.
−20°C
SMILES string
[Br-].O=C1CCC(=O)N1OC(=O)c2cccc(C[n+]3ccc(cc3)-c4cnc(o4)-c5ccc6OCCCc6c5)c2
InChI
1S/C29H24N3O6.BrH/c33-26-8-9-27(34)32(26)38-29(35)23-4-1-3-19(15-23)18-31-12-10-20(11-13-31)25-17-30-28(37-25)22-6-7-24-21(16-22)5-2-14-36-24;/h1,3-4,6-7,10-13,15-17H,2,5,8-9,14,18H2;1H/q+1;/p-1
Inchi Key
LSKYMYVQAVWWBK-UHFFFAOYSA-M
General description
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service