Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52318

Sigma-Aldrich

n-Hexadecyl β-D-maltoside

≥99.0% (TLC)

Synonym(s):

Cetyl β-D-maltoside, Hexadecyl-β-D-maltoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H54O11
CAS Number:
Molecular Weight:
566.72
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

assay

≥99.0% (TLC)

form

solid

optical activity

[α]20/D +45±2°

mol wt

566.72 g/mol

CMC

0.0006 mM (20-25°C)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

InChI

1S/C28H54O11/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-36-27-25(35)23(33)26(20(18-30)38-27)39-28-24(34)22(32)21(31)19(17-29)37-28/h19-35H,2-18H2,1H3/t19-,20-,21-,22+,23-,24-,25-,26-,27-,28-/m1/s1

InChI key

GTQCHJYVKDXMRU-YMEALESQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Detergent for the purification, extraction and solubilization of membrane-bound proteins in the native structure. It increases solubilization by dissociating aggregates and is easily removed by dialysis: N.C. Robinson et al., Biochemistry, 24, 6298 (1985)

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service