Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51878

Sigma-Aldrich

α-Ketosuberic acid

≥97.0% (HPLC)

Synonym(s):

2-Ketosuberic acid, 2-Oxooctanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O5
CAS Number:
Molecular Weight:
188.18
Beilstein/REAXYS Number:
1778635
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

assay

≥97.0% (HPLC)

form

solid

storage temp.

2-8°C

InChI

1S/C8H12O5/c9-6(8(12)13)4-2-1-3-5-7(10)11/h1-5H2,(H,10,11)(H,12,13)

InChI key

HAGOOZVJLSSXGZ-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Randy M Drevland et al.
The Journal of biological chemistry, 283(43), 28888-28896 (2008-09-04)
Homoaconitase enzymes catalyze hydrolyase reactions in the alpha-aminoadipate pathway for lysine biosynthesis or the 2-oxosuberate pathway for methanogenic coenzyme B biosynthesis. Despite the homology of this iron-sulfur protein to aconitase, previously studied homoaconitases catalyze only the hydration of cis-homoaconitate to
David E Graham
Methods in enzymology, 494, 301-326 (2011-03-16)
Coenzyme M (CoM) and coenzyme B (CoB) are essential for methane production by the euryarchaea that employ this specialized anaerobic metabolism. Two pathways are known to produce CoM, 2-mercaptoethanesulfonate, and both converge on the 2-oxoacid sulfopyruvate. These cells have recruited
Yuchen Liu et al.
Environmental microbiology, 14(10), 2632-2644 (2012-05-26)
Studies on sulfur metabolism in archaea have revealed many novel enzymes and pathways and have advanced our understanding on metabolic processes, not only of the archaea, but of biology in general. A variety of dissimilatory sulfur metabolisms, i.e. reactions used
R H White
Biochemistry, 28(24), 9417-9423 (1989-11-28)
The biosynthetic steps involved in the conversion of alpha-ketosuberate to 7-mercaptoheptanoic acid were studied in cell-free extracts of methanogenic bacteria. The pathway was established by measuring the incorporation of stable isotopically labeled precursors into the S-methyl ether methyl ester derivative
Jeyaraman Jeyakanthan et al.
Biochemistry, 49(12), 2687-2696 (2010-02-23)
The aconitase family of hydro-lyase enzymes includes three classes of proteins that catalyze the isomerization of alpha-hydroxy acids to beta-hydroxy acids. Besides aconitase, isopropylmalate isomerase (IPMI) proteins specifically catalyze the isomerization of alpha,beta-dicarboxylates with hydrophobic gamma-chain groups, and homoaconitase (HACN)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service