Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51821

Sigma-Aldrich

Fluorescent red 751 reactive

suitable for fluorescence, ≥90% (coupling to amines)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H54N3NaO11S2
Molecular Weight:
912.05
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

assay

≥90% (coupling to amines)

fluorescence

λex 660; λem 765 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

Storage temp.

−20°C

SMILES string

[Na+].CCN1c2cc3[o+]c(cc(\C=C\C=C\C=C4/N(CCCS([O-])(=O)=O)c5ccc(cc5C4(C)CCCC(=O)ON6C(=O)CCC6=O)S([O-])(=O)=O)c3cc2C(C)=CC1(C)C)C(C)(C)C

InChI

1S/C46H55N3O11S2.Na/c1-9-48-37-28-38-34(27-33(37)30(2)29-45(48,6)7)31(25-40(59-38)44(3,4)5)15-11-10-12-16-39-46(8,22-13-17-43(52)60-49-41(50)20-21-42(49)51)35-26-32(62(56,57)58)18-19-36(35)47(39)23-14-24-61(53,54)55;/h10-12,15-16,18-19,25-29H,9,13-14,17,20-24H2,1-8H3,(H-,53,54,55,56,57,58);/q;+1/p-1/b12-10+,15-11+,39-16-;

Inchi Key

WXZMYCKLBDQDAE-ZRLYFAETSA-M

General description

Fluorescent red 751 reactive is a new fluorescent label for the near infrared area. Extremely long wavelength fluorescence is especially well suited for applications where sample autofluorescence might be critical or penetration of tissue or other matrix is required. Fluorescent red 751 reactive shows strong fluorescence (molar absorption 220 000 M-1cm-1)

Application

Fluorescent red 751 reactive is an amine conjugating fluorescent biolabel that can be used for covalent coupling to proteins and other biomolecules containing primary amino groups like amino modified DNA-oligomers and amino modified biotin. Fluorescent red 751 reactive is a new fluorescent label for the near infrared area. Extremely long wavelength fluorescence is especially well suited for applications where sample autofluorescence might be critical or penetration of tissue or other matrix is required.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service