Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51796

Sigma-Aldrich

Atto-Mono 2

BioReagent

Synonym(s):

Nucleic acid stain II solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500

product line

BioReagent

form

liquid

manufacturer/tradename

ATTO-TEC GmbH

concentration

2.0 mM (UV/Vis)

suitability

passes test for fluorescence

storage temp.

−20°C

Analysis Note

Atto-Mono 2 is a cell permeable monomer. In aqeous solution it does not show significant fluorescence. In presence of DNA it shows a bright fluorescence with quantum yield above 25%. Maximum of absorption lies at 515 nm, exctinction coefficient 68500 M-1cm-1. Maximum of emission shifts from 538 nm to 610 nm if bound to DNA.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service