51653C
Ham′s Nutrient Mixture F12
liquid
Synonym(s):
Nutrient Mixture F12 Ham’s
About This Item
Recommended Products
description
for research or for further manufacturing use
without L-glutamine, without hypoxanthine
form
liquid
quality
for further manufacturing use
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service