Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51306

Sigma-Aldrich

TRIS glycine SDS buffer solution

BioReagent, for molecular biology, 10 × in concentrate

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41105319
eCl@ss:
32129211
NACRES:
NA.25

grade

Molecular Biology
for molecular biology

product line

BioReagent

form

liquid

concentration

10 × in concentrate

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

pH

8.6±0.2

density

1.07 g/mL at 20 °C

Looking for similar products? Visit Product Comparison Guide

Physical form

0.25 M Tris, 1.92 M Glycine and 0.1% SDS

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurélie Morin et al.
Cell reports, 30(13), 4551-4566 (2020-04-03)
Loss-of-function mutations in the SDHB subunit of succinate dehydrogenase predispose patients to aggressive tumors characterized by pseudohypoxic and hypermethylator phenotypes. The mechanisms leading to DNA hypermethylation and its contribution to SDH-deficient cancers remain undemonstrated. We examine the genome-wide distribution of 5-methylcytosine and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service