Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51304

Sigma-Aldrich

Ampholyte high resolution

pH 3.0-6.0, ~40% in H2O

Synonym(s):

SinuLyte

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.26
Product 51304 is not currently sold in your country. Contact Technical Service

concentration

~40% in H2O

technique(s)

electrophoresis: suitable

pH

3.0-6.0

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This series of carrier ampholytes is based on a specific method to produce mixtures of numerous polyamino-polycarboxylic acids. These carrier ampholytes have been developed for use of isoelectric focusing in quality control applications, especially in food and seed analysis.
  • High buffer capacity: Compared to established ampholytes they offer buffering capacities 1.5-2.5 times higher (up to pH 8, equal above pH 8)
  • Good, even conductivity
  • Linear and reproducible pH gradient
  • High resolution

Legal Information

SinuLyte is a trademark of Sinus Biochemistry and Electrophoresis GmbH

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service