Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51071

Sigma-Aldrich

Guanosine 5′-[β,γ-imido]triphosphate trisodium salt hydrate

~80% (HPLC)

Synonym(s):

β:γ-Imidoguanosine 5′-triphosphate trisodium salt hydrate, 5′-Guanylyl-imidodiphosphate trisodium salt hydrate, GMP-PNP, Gpp(NH)p

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N6Na3O13P3 · xH2O
CAS Number:
Molecular Weight:
588.14 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

assay

~80% (HPLC)

impurities

≤3% solvent (ethanol, methanol)

solubility

H2O: 50 mg/mL, clear, colorless

shipped in

dry ice

storage temp.

−70°C

SMILES string

[Na+].[Na+].[Na+].Nc1nc(O)c2ncn([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)NP(O)([O-])=O)[C@@H](O)[C@H]3O)c2n1

InChI

1S/C10H17N6O13P3.3Na/c11-10-13-7-4(8(19)14-10)12-2-16(7)9-6(18)5(17)3(28-9)1-27-32(25,26)29-31(23,24)15-30(20,21)22;;;/h2-3,5-6,9,17-18H,1H2,(H,25,26)(H3,11,13,14,19)(H4,15,20,21,22,23,24);;;/q;3*+1/p-3/t3-,5-,6-,9-;;;/m1.../s1

InChI key

QGJNRMUNXAROIT-CYCLDIHTSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Non-hydrolyzable analog of GTP; the preferred GTP analog to activate ADP-ribosylation factor.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frank Adolf et al.
Cell reports, 26(1), 250-265 (2019-01-04)
Intracellular transport and homeostasis of the endomembrane system in eukaryotic cells depend on the formation and fusion of vesicular carriers. Coat protein complex (COP) II vesicles export newly synthesized secretory proteins from the endoplasmic reticulum (ER), whereas COPI vesicles facilitate traffic
Sebastian Daum et al.
Biological chemistry, 395(7-8), 801-812 (2014-07-09)
As shape transformations of membranes are vital for intracellular trafficking, it is crucial to understand both the mechanics and the biochemistry of these processes. The interplay of these two factors constitutes an experimental challenge, however, because biochemical experiments are not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service