Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50788

Sigma-Aldrich

2,5-Hexanediol 3 M solution

Synonym(s):

2,5-Hexanediol solution, Additive Screening Solution 06/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

CC(O)CCC(C)O

InChI

1S/C6H14O2/c1-5(7)3-4-6(2)8/h5-8H,3-4H2,1-2H3

InChI key

OHMBHFSEKCCCBW-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Nylén et al.
Archives of toxicology, 67(6), 435-441 (1993-01-01)
Albino (Sprague-Dawley) and pigmented (Norwegian Brown) male rats were exposed to 2,5-hexanediol (H; 1%) in their drinking water for 5 or 8 weeks, respectively. Half of the rats of each strain were housed in light (average 30 cd/cm2 inside cage
Lv Li et al.
Brain research, 1159, 8-17 (2007-06-19)
Opposing functions of activated microglia, namely neuroprotection or neurotrophy versus neurodestruction or neurotoxicity, have been observed in a number of experimental models of neurotrauma and neurodegenerative diseases. However, the mechanism(s) involved in the determination of which function activated microglia execute
J B Cavanagh
Neuropathology and applied neurobiology, 8(1), 19-34 (1982-01-01)
Rats have been dosed with 2,5-hexanediol for 48 days and then allowed to recover. The changes in the accumulations of neurofilamentous masses in various pathways in CNS and PNS have been followed by light microscopy over the subsequent 9 weeks.
Interaction between neurotoxicities induced by organophosphorus and long-chain hexacarbon compounds.
M B Abou-Donia
Neurotoxicology, 4(4), 117-135 (1983-01-01)
A K Agrawal et al.
Neurotoxicology, 6(3), 53-59 (1985-01-01)
Single exposure of 150, 300 and 600 mg/kg 2,5-hexanediol, (2,5 HD) a metabolite of n-hexane showed no significant effect on 3H-spiroperidol binding to corpus striatal membranes when compared with control animals, where as repeated exposure of 2,5-HD, 300 and 600

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service