Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50241

Sigma-Aldrich

Lithium chloride 20 mM – Cobalt(II) chloride 5 mM – Sodium cacodylate Buffer pH 6.0; 50 mM – Dioxan 20% solution

Synonym(s):

Crystallization solution 45/Kit-No 18839

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

pH

6.0±0.2 ( in neat)

density

1.02 g/mL at 20 °C

storage temp.

2-8°C

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

target_organs

Respiratory system

supp_hazards

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service