49328
EVOblue™ 30
≥90.0% (coupling to amines)
Synonym(s):
3-[N-(2-Carboxyethyl)methylamino]-7-[N-ethyl(3-sulfonatopropyl)amino]phenoxazin-5-ium
About This Item
assay
≥90.0% (coupling to amines)
solubility
DMF: soluble
fluorescence
λex 635 nm; λem 666 nm in 0.1 M phosphate pH 7.0
suitability
suitable for fluorescence
Storage temp.
−20°C
SMILES string
CCN(CCCS([O-])(=O)=O)c1ccc2nc3ccc(cc3[o+]c2c1)N(C)CCC(O)=O
InChI
1S/C21H25N3O6S/c1-3-24(10-4-12-31(27,28)29)16-6-8-18-20(14-16)30-19-13-15(5-7-17(19)22-18)23(2)11-9-21(25)26/h5-8,13-14H,3-4,9-12H2,1-2H3,(H-,25,26,27,28,29)
Inchi Key
RCTKCVWZEGPPCK-UHFFFAOYSA-N
Application
Features and Benefits
Other Notes
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service