Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

492-05F

Sigma-Aldrich

Human Marrow Stromal Cells: HMSC, fetal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human bone marrow

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 46

morphology

stromal

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

tuberculosis

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Bone marrow is the major blood creating organ, but in addition to supporting hematopoietic growth and differentiation, marrow stromal cells can be induced to produce cells of other connective tissues, such as bone, cartilage, and fat, as well as cells from neuroectodermal (neurons) and endodermal (hepatocytes) lineages. The potential of HMSCs to maintain multipotency and proliferate extensively in vitro provides new avenues for cell-based therapy in the restoration of damaged or diseased tissue.

HMSC have been utilized to investigate the effects of TGF-β1 on differentiation toward different lineages, and show that it inhibits adipogenic differentiation and induces either chondrogenic, osteogenic or smooth muscle cell differentiation, depending the specific culture conditions (Zhao, 2011); to develop methods to assess differentiation potential of stem cells without the differentiation induction (Hirota, 2011); to compare HLA expression profiles of bone marrow-derived and adipose-derived stem cells (Purandare, 2013); and to design heart valve-shaped tri-layered collagen-based constructs for stem cell seeding and differentiation to use in heart valve tissue engineering (Tedder, 2010, 2011).

Cell Line Origin

Bone Marrow

Application

blood production, hematopoietic growth and differentiation, pathways of pluripotent cell differentiation into bone, cartilage and fat, connective tissue production,supportive structure, cell production, cell proliferation, cell based therapy, tissue restoration and repair, differentiation, effects of culture conditinos, HLA expression, cytokine production, immunomodulation, clinical trials

Components

Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 10 doublings

Subculture Routine

Please refer to the HMSC Culture Protocol.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service