49071
3′-Sialyl-N-acetyllactosamine
Na salt, ≥90% (TLC)
About This Item
Recommended Products
biological source
synthetic
Quality Level
assay
≥90% (TLC)
form
crystalline
storage temp.
−20°C
SMILES string
CC(=O)NC1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O[C@]3(C[C@H](O)[C@@H](NC(C)=O)[C@@H](O3)[C@H](O)[C@H](O)CO)C(O)=O)[C@H]2O)[C@H](O)[C@H]1O
InChI
1S/C25H42N2O19/c1-7(31)26-13-9(33)3-25(24(40)41,45-20(13)14(35)10(34)4-28)46-21-15(36)11(5-29)43-23(18(21)39)44-19-12(6-30)42-22(27-8(2)32)17(38)16(19)37/h9-23,28-30,33-39H,3-6H2,1-2H3,(H,26,31)(H,27,32)(H,40,41)/t9-,10+,11+,12+,13+,14+,15-,16+,17+,18+,19+,20+,21-,22?,23-,25+/m0/s1
InChI key
HUNVSYPDCXFGLB-KROWHOKKSA-N
Application
Other Notes
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service