Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46049

Supelco

Diniconazole

PESTANAL®, analytical standard

Synonym(s):

α-tert-Butyl-β-(2,4-dichlorophenylmethylene)-1H-1,2,4-triazole-1-ethanol, Diclopentezol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17Cl2N3O
CAS Number:
Molecular Weight:
326.22
Beilstein/REAXYS Number:
9202976
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Product 46049 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

description

mixture of Z and E isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)(C)C(O)\C(=C\c1ccc(Cl)cc1Cl)n2cncn2

InChI

1S/C15H17Cl2N3O/c1-15(2,3)14(21)13(20-9-18-8-19-20)6-10-4-5-11(16)7-12(10)17/h4-9,14,21H,1-3H3/b13-6-

InChI key

FBOUIAKEJMZPQG-MLPAPPSSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diniconazole belongs to the class of triazole fungicides used to control spoilage of crops and maintain the quality of agricultural products.[1]

Application

Diniconazole may be used as an analytical reference standard for the quantification of the analyte in vegetable samples, biological samples and earth worms using different chromatography techniques.[2][1]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>302.0 °F

flash_point_c

> 150 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LC-MS/MS method for simultaneous determination of myclobutanil, hexaconazole, diniconazole, epoxiconazole and tetraconazole enantiomers in soil and earthworms
International Journal of Environmental Analytical Chemistry, 94(8), 791-800 (2014)
Determination of tetraconazole and diniconazole fungicide residues in tomatoes and green beans by capillary gas chromatography
Amer.MM, et al.
Yakugaku Zasshi, 127(6), 993-999 (2007)
Determination of triazole fungicides in environmental water samples by high performance liquid chromatography with cloud point extraction using polyethylene glycol 600 monooleate
Tang T, et al.
Analytica Chimica Acta, 680(1-2), 26-31 (2010)
Zhenjiang Liu et al.
PloS one, 7(10), e46929-e46929 (2012-10-12)
In the European Union (EU), the use of diniconazole-M is no longer authorized. However, residues of diniconazole-M occur in various plant commodities. A selective and simple analytical method for the trace level determination of diniconazole in soil, fruit, vegetables and
Yinggao Liu et al.
Journal of experimental botany, 61(11), 2979-2990 (2010-05-13)
H(2)O(2) is known as a signal molecule in plant cells, but its role in the regulation of aqbscisic acid (ABA) and gibberellic acid (GA) metabolism and hormonal balance is not yet clear. In this study it was found that H(2)O(2)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service