Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45167

Sigma-Aldrich

Endoproteinase Pro-C, recombinant from E. coli

~3 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

form

lyophilized solid

specific activity

~3 U/mg

analyte chemical class(es)

amino acids

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Endoproteinase Pro-C cleaves peptide bonds of non-terminal amino acids following a proline residue.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol p-nitroaniline from Z-Gly-Pro-pNA per minute at pH 7.0 and 25°C

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endoproteinase Pro-C-Catalyzed Peptide Bond Formation in Frozen Aqueous Systems
M. Haensler, H.-D. Jakubke
Enz. Microbiol. Technol., 22, 617-620 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service