Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44688

Sigma-Aldrich

2-(2-Naphthyl)-2-(pentyloxy)acetonitrile

≥95% (HPLC)

Synonym(s):

α-Pentyloxy-2-naphthaleneacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO
CAS Number:
Molecular Weight:
253.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥95% (HPLC)

form

liquid

fluorescence

λex 355 nm; λem 368 nm in methanol

SMILES string

CCCCCOC(C#N)c1ccc2ccccc2c1

InChI

1S/C17H19NO/c1-2-3-6-11-19-17(13-18)16-10-9-14-7-4-5-8-15(14)12-16/h4-5,7-10,12,17H,2-3,6,11H2,1H3

Inchi Key

AFNOPYUTGOKTHI-UHFFFAOYSA-N

Pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service