Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44673

SAFC

2-(Benzyloxy)-2-(2-naphthyl)acetonitrile

≥95.0%

Synonym(s):

α-(Benzyloxy)-2-naphthaleneacetonitrile, 2-(2-Naphthyl)-2-(phenylmethoxy)acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15NO
CAS Number:
Molecular Weight:
273.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥95.0%

fluorescence

λex 227 nm; λem 336 nm in methanol

SMILES string

N#CC(OCc1ccccc1)c2ccc3ccccc3c2

InChI

1S/C19H15NO/c20-13-19(21-14-15-6-2-1-3-7-15)18-11-10-16-8-4-5-9-17(16)12-18/h1-12,19H,14H2

Inchi Key

SLMXQIWKVZNVSR-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service