Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44627

Sigma-Aldrich

D-Fructopyranose 2-phosphate potassium salt

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13O9P · xK+
CAS Number:
Molecular Weight:
260.14 (free acid basis)
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

assay

≥98.0% (TLC)

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](O)[C@H](O)CO[C@]1(OP(O)(O)=O)CO

InChI

1S/C6H13O9P/c7-2-6(15-16(11,12)13)5(10)4(9)3(8)1-14-6/h3-5,7-10H,1-2H2,(H2,11,12,13)/t3-,4?,5-,6+/m1/s1

InChI key

FKTOSTOKRBVXKU-CNKSGKBASA-N

Biochem/physiol Actions

Metabolite involved in the fructose and mannose system.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of glycosyl phosphates. Betad-fructopyranose 2-phosphate.
D L MacDonald
The Journal of organic chemistry, 31(2), 513-516 (1966-02-01)
C Degani et al.
Journal of molecular evolution, 6(1), 51-60 (1975-10-03)
Cyanogen-induced phosphorylation of D-fructose at pH 8.8 led to the formation of a phosphorylated sugar identified as alpha-D-furcto-pyranose 2-phosphate on the basis of its chromatographic and electrophoretic properties, its lability to hydrolysis by alkaline phosphatase, the rate of its acid-catalysed
Hydrolysis and Isomerization of Sugar Phosphates and Carbohydrate Phosphodiesters.
Mikkola, S.
Current Organic Chemistry, 17, 1525-1544 (2013)
Properties of D-fructose 1,2-cyclic phosphates and fructose 2-phosphates.
Taniguchi, H. and Nakamura, M.
Agricultural and Biological Chemistry, 36, 2373-2380 (1972)
SYNTHESIS OF D-FRUCTOPYRANOSE 2-PHOSPHATE AND D-FRUCTOFURANOSE 2-PHOSPHATE.
H G PONTIS et al.
The Biochemical journal, 89, 452-459 (1963-12-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service