Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44049

Sigma-Aldrich

11-Aminoundecanoic acid tetramethylammonium salt

≥97.0% (NT)

Synonym(s):

Tetramethylammonium 11-aminoundecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H34N2O2
CAS Number:
Molecular Weight:
274.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥97.0% (NT)

SMILES string

C[N+](C)(C)C.NCCCCCCCCCCC([O-])=O

InChI

1S/C11H23NO2.C4H12N/c12-10-8-6-4-2-1-3-5-7-9-11(13)14;1-5(2,3)4/h1-10,12H2,(H,13,14);1-4H3/q;+1/p-1

InChI key

VREIIGDEUQSHDP-UHFFFAOYSA-M

Application

11-Aminoundecanoic acid tetramethylammonium salt may be used as a bipolar biocompatible surfactant in the development of nanoparticles for use in drug delivery, magnetic resonance imaging and magnetic hyperthermia.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiyan Yan et al.
Journal of nanoscience and nanotechnology, 5(8), 1185-1192 (2005-10-01)
The therapeutic effect of Fe2O3 nanoparticles combined with magnetic fluid hyperthermia (MFH) on human hepatocarcinoma SMMC-7721 cells in vitro and xenograft liver cancer in nude mice is studied. We examined growth and apoptosis of SMMC-7721 cells treated with MFH containing
Zi-Yu Wang et al.
World journal of gastroenterology, 15(24), 2995-3002 (2009-06-26)
To study the methods of preparing the magnetic nano-microspheres of Fe(2)O(3) and As(2)O(3)/Fe(2)O(3) complexes and their therapeutic effects with magnetic fluid hyperthermia (MFH). Nanospheres were prepared by chemical co-precipitation and their shape and diameter were observed. Hemolysis, micronucleus, cell viability
B Ankamwar et al.
Nanotechnology, 21(7), 75102-75102 (2010-01-22)
In order to reveal the biocompatibility of Fe(3)O(4) nanoparticles and bipolar surfactant tetramethylammonium 11-aminoundecanoate cytotoxicity tests were performed as a function of concentration from low (0.1 microg ml(-1)) to higher concentration (100 microg ml(-1)) using various human glia, human breast

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service