43683
Disialoganglioside GD3 from bovine milk
≥90% (TLC), ammonium salt
Synonym(s):
Ganglioside GD3, disialo
About This Item
biological source
microbial
assay
≥90% (TLC)
form
powder
lipid type
sphingolipids
storage temp.
−20°C
Biochem/physiol Actions
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
No data available
flash_point_c
No data available
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service