Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43413

Sigma-Aldrich

Anti-Rabbit IgG - Tracy 652 from goat antiserum

1 mg/mL protein

Synonym(s):

Tracy 652 - goat-anti-rabbit IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

Tracy 652 conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

polyclonal

contains

50% glycerol as stabilizer

species reactivity

rabbit

concentration

1 mg/mL protein

fluorescence

λex 652 nm; λem 672 nm in PBS

storage temp.

−20°C

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections Tracy 652 is a newly developed highly water-soluble and stable NIR fluorescent Dye with spectral range above 650 nm is of particular interest in biology analysis and imaging due to the reduced background and improved sensitivity
Anti-Rabbit IgG - Tracy 652 is specific for rabbit IgG subclasses.

Immunogen

rabbit IgG

Application

Anti-Rabbit IgG - Tracy 652 may be used in fluorescence-based assays.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

unconjugated dye ≤5% of total fluorescence; dye-to-protein ratio >2

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Placental transfer of IgG subclasses in a Japanese population
Hashira S et al
Pediatrics International : Official Journal of the Japan Pediatric Society, 45, 337-342 (2000)
Qing Zhang et al.
Molecular medicine reports, 11(6), 4685-4693 (2015-02-13)
Ovarian cancer is a life‑threatening disease in females worldwide. The polarization of macrophages is crucial in oncogenesis and the development of ovarian cancer. Increasing evidence has supported the correlation between ovarian cancer stem‑like cells (OCSCs) and macrophages, however, whether OCSCs
Xue Zhou et al.
Molecular medicine reports, 15(4), 2204-2212 (2017-03-06)
The commonly used inhalation anesthetic, sevoflurane, has been previously demonstrated to induce apoptosis in the developing brain; however, the underlying molecular mechanisms remain largely unknown. MicroRNAs (miRNAs) serve important roles in multiple physiological/pathological processes, such as cell death and survival.
Weijuan Huang et al.
Molecular medicine reports, 11(2), 805-814 (2014-11-07)
It has been previously demonstrated that Astragalus and Paeoniae radix rubra extract (APE) had a protective effect against liver fibrosis in mice. The present study aimed to investigate the hepatoprotective effect of APE on CCl4‑induced hepatic fibrosis in rats. Liver
Sen Yang et al.
Journal of dairy science, 103(4), 2993-3001 (2020-02-15)
Bovine β-lactoglobulin (β-LG) is the major allergen in milk powder. The IgG/IgE binding capacity and structural characteristics of β-LG after spray drying in the presence or absence of α-lactose at 120 and 180°C were investigated by ELISA and mass spectrometry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service