Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4339039

Sigma-Aldrich

Cleavable iCAT® Reagent Bulk Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

General description

Contains 100 units of both Heavy and Light reagent, Cleaving Reagent A, and operating instructions. Each unit of reagent can label up to ~100 μg of protein.

Analysis Note

To view the Protocol for the Cleavable ICAT Reagent Kit for Protein Labeling (monoplex version), please visit this Protocol link

To view the Protocol for the 1-D PAGE Cleavable ICAT Reagent Applications Development Kit for Targeted Protein ID and Quantitation (monoplex version), please visit this Protocol link.

To view the Quick Reference Procedure for the 1-D PAGE Cleavable ICAT Reagent Applications Development Kit for Targeted Protein ID and Quantitation (monoplex version), please visit this Quick Reference Procedure link.

To view the Quick Reference Card for the Cleavable ICAT Reagent Kit for Protein Labeling, please visit this Quick Reference Card link.

Legal Information

iCAT is a registered trademark of University of Washington

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service