Key Documents
4339036
Sigma-Aldrich
Cleavable iCAT® Reagent - 10 Assay Kit
About This Item
UNSPSC Code:
12352200
storage temp.
2-8°C
General description
Cleavable ICAT Reagents 10 Assay Kit contains 10 units of Heavy reagent and 11 units of Light reagent. Each unit of reagent can label up to ~100 μg of protein.
Analysis Note
To view the Protocol for the Cleavable ICAT Reagent Kit for Protein Labeling (monoplex version), please visit this Protocol link
To view the Protocol for the 1-D PAGE Cleavable ICAT Reagent Applications Development Kit for Targeted Protein ID and Quantitation (monoplex version), please visit this Protocol link.
To view the Quick Reference Procedure for the 1-D PAGE Cleavable ICAT Reagent Applications Development Kit for Targeted Protein ID and Quantitation (monoplex version), please visit this Quick Reference Procedure link.
To view the Quick Reference Card for the Cleavable ICAT Reagent Kit for Protein Labeling, please visit this Quick Reference Card link.
Legal Information
iCAT is a registered trademark of University of Washington
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 1
supp_hazards
Storage Class
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Trairak Pisitkun et al.
Physiological genomics, 25(2), 263-276 (2006-02-02)
The inner medullary collecting duct (IMCD) is an important site of vasopressin-regulated water and urea transport. Here we have used protein mass spectrometry to investigate the proteome of the IMCD cell and how it is altered in response to long-term