Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43381

Sigma-Aldrich

(S)-Sodium 2,3-dihydroxyisovalerate hydrate

≥95.0% (CE)

Synonym(s):

(S)-2,3-Dihydroxy-3-methylbutanoic acid sodium salt, L(+)-α,β-Dihydroxyisovaleric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O4
Molecular Weight:
134.13
Beilstein/REAXYS Number:
4657323
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

assay

≥95.0% (CE)

optical purity

enantiomeric excess: ≥98.0%

storage temp.

2-8°C

SMILES string

O.[Na+].CC(C)(O)[C@H](O)C([O-])=O

InChI

1S/C5H10O4.Na.H2O/c1-5(2,9)3(6)4(7)8;;/h3,6,9H,1-2H3,(H,7,8);;1H2/q;+1;/p-1/t3-;;/m1../s1

InChI key

KGRRZNPTTZIUMS-HWYNEVGZSA-M

Application

Sodium 2,3-dihydroxyisovalerate hydrate may be used to study the distribution, function and kinetics of dihyroxy-acid dehydratases such as the superoxide-sensitive enzyme α, β-dihydroxyisovalerate dehydratase.

Biochem/physiol Actions

Metabolite in the pantothenate and CoA biosynthesis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C F Kuo et al.
The Journal of biological chemistry, 262(10), 4724-4727 (1987-04-05)
Increasing the intracellular flux of O-2 by incubating aerobic Escherichia coli with paraquat or plumbagin markedly lowered the alpha, beta-dihydroxyisovalerate dehydratase activity detectable in extracts from these cells. This effect was not seen in the absence of dioxygen and was
O R Brown et al.
Archives of biochemistry and biophysics, 319(1), 10-22 (1995-05-10)
Dihydroxy-acid dehydratase (DHAD) has a [4Fe-4S] cluster and is reported to be facilely inactivated by oxidant stress. To directly assess the biological effects in vivo of superoxide dismutase (SOD) on the oxidant sensitivity of DHAD, we used an Escherichia coli
Concepts and perspectives in enzyme stereochemistry.
K R Hanson
Annual review of biochemistry, 45, 307-330 (1976-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service