Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43187

Sigma-Aldrich

Nano beads based on PD, Chromeon 470-marked

Synonym(s):

PD-Chromeon 470

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

fluorescence

λex 470 nm; λem 603 nm in H2O

storage temp.

2-8°C

Features and Benefits

PD is a new polymer, which provides less sensitivity to oxygen and reduced photo bleaching compared to latex (polystyrene) particles. PD particles are ideally suited for phosphorescent lifetime (decay time) measurements. Fluorescent PD particles are suitable for protein labeling and detection of antigen-antibody reactions.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service