Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43184

Sigma-Aldrich

Monopotassium 7-amino-1,3-naphthalenedisulfonate hydrate

suitable for fluorescence, ≥98.0% (HPLC)

Synonym(s):

2-Naphthylamine-6,8-disulfonic acid monopotassium salt hydrate, 7-Amino-1,3-naphthalenedisulfonic acid monopotassium salt hydrate, AGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H5(SO3H)SO3K · xH2O
CAS Number:
Molecular Weight:
302.30 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

assay

≥98.0% (HPLC)

impurities

≤8.0% water

mp

>300 °C (lit.)

suitability

suitable for fluorescence

SMILES string

[K+].[H]O[H].Nc1ccc2cc(cc(c2c1)S(O)(=O)=O)S([O-])(=O)=O

InChI

1S/C10H9NO6S2.K.H2O/c11-7-2-1-6-3-8(18(12,13)14)5-10(9(6)4-7)19(15,16)17;;/h1-5H,11H2,(H,12,13,14)(H,15,16,17);;1H2/q;+1;/p-1

InChI key

SHURQDDWGCOLFA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Monopotassium 7-amino-1,3-naphthalenedisulfonate may be used to conjugate the reducing ends of oligosaccharides, such as those derived from glycoproteins, glycolipids, and proteoglycans, for analysis via gel electrophoresis.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K B Lee et al.
Carbohydrate research, 214(1), 155-168 (1991-07-18)
A new method is described for sequencing linear oligosaccharides on gels using charged, fluorescent conjugates. The reducing ends of various mono-, di-, tri-, and tetra-saccharides were conjugated with monopotassium 7-amino-1,3-naphthalenedisulfonate (a fluorescent and negatively charged compound) by reductive amination using
Rafael Parra-Medina et al.
PloS one, 10(5), e0124040-e0124040 (2015-05-06)
Celiac disease (CD) is an immune-mediated enteropathy triggered by the ingestion of gluten in susceptible individuals, and its prevalence varies depending on the studied population. Given that information on CD in Latin America is scarce, we aimed to investigate the
Mingyan Hei et al.
American journal of perinatology, 32(8), 725-732 (2014-12-24)
This study aimed to compare the outcomes for symmetrical (sSGA) versus asymmetrical (aSGA) small for gestational age (SGA) < 32 weeks preterm infants. A total of 12,179 eligible infants admitted to the Canadian tertiary neonatal intensive care units (NICUs) over a 7-year-period
David R Holmes et al.
Journal of the American College of Cardiology, 65(24), 2614-2623 (2015-06-20)
The risk-benefit ratio of left atrial appendage closure (LAAC) versus systemic therapy (warfarin) for prevention of stroke, systemic embolism, and cardiovascular death in nonvalvular atrial fibrillation (NVAF) requires continued evaluation. This study sought to assess composite data regarding left atrial
Ben J Evans et al.
Cytogenetic and genome research, 145(3-4), 243-252 (2015-06-13)
Genome duplication creates redundancy in proteins and their interaction networks, and subsequent smaller-scale gene duplication can further amplify genetic redundancy. Mutations then lead to the loss, maintenance or functional divergence of duplicated genes. Genome duplication occurred many times in African

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service