43095
Diphenylmaleic anhydride
suitable for fluorescence, ≥98.0% (HPLC)
Synonym(s):
3,4-Diphenyl-2,5-furandione
About This Item
Recommended Products
assay
≥98.0% (HPLC)
mp
155-159 °C
solubility
DMF: soluble
fluorescence
λex 284 nm; λem 506 nm (after derivatization with hexylamine)
suitability
suitable for fluorescence
SMILES string
O=C1OC(=O)C(c2ccccc2)=C1c3ccccc3
InChI
1S/C16H10O3/c17-15-13(11-7-3-1-4-8-11)14(16(18)19-15)12-9-5-2-6-10-12/h1-10H
InChI key
OUJCFCNZIUTYBH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Other Notes
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service