43048
Atto 740-Streptavidin
suitable for fluorescence
Synonym(s):
Streptavidin-Atto 740
About This Item
Recommended Products
manufacturer/tradename
ATTO-TEC GmbH
solubility
DMF: 1 mg/mL, clear
fluorescence
λex 692 nm; λem 764 nm±5 nm
suitability
corresponds for SDS-PAGE
suitable for fluorescence
storage temp.
−20°C
General description
Atto 740 is a pH sensitive product. While practically stable up to pH 7.4 (PBS-buffer), it slowly degrades at higher pH. If exposed to higher pH for coupling purposes, we recommend reducing the pH immediately after completion of the reaction.
find more information here
Packaging
Analysis Note
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service