Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42932

Sigma-Aldrich

N-[4-(Aminomethyl)benzyl]rhodamine 6G-amide bis(trifluoroacetate)

suitable for fluorescence, BioReagent, ≥95.0% (HPCE)

Synonym(s):

Rhodamine 6G-N-[4-(aminomethyl)benzyl]amide bis(trifluoroacetate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H36N4O2·C4H2F6O4
CAS Number:
Molecular Weight:
760.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

product line

BioReagent

Quality Level

assay

≥95.0% (HPCE)

fluorescence

λex 528 nm; λem 553 nm in 0.1 M phosphate pH 3.0

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.CCNc1cc2OC3=CC(=N\CC)\C(C)=CC3=C(c2cc1C)c4ccccc4C(=O)NCc5ccc(CN)cc5

InChI

1S/C34H36N4O2.2C2HF3O2/c1-5-36-29-17-31-27(15-21(29)3)33(28-16-22(4)30(37-6-2)18-32(28)40-31)25-9-7-8-10-26(25)34(39)38-20-24-13-11-23(19-35)12-14-24;2*3-2(4,5)1(6)7/h7-18,36H,5-6,19-20,35H2,1-4H3,(H,38,39);2*(H,6,7)/b37-30-;;

InChI key

NWLZLJKDDQUWKR-FXIQUCANSA-N

Application

N-[4-(Aminomethyl)benzyl]rhodamine 6G-amide bis(trifluoroacetate) is a rhodamine 6G-amide derivative useful as a bioreagent in fluorescence assays.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Yuan et al.
Organic & biomolecular chemistry, 9(6), 1723-1726 (2011-01-25)
A novel strategy to systematically tune the pK(a) values of rhodamines is described. This strategy was applied to rationally develop compound 1e with a pK(a) of 6.5, the highest among rhodamine amide derivatives, and it could be employed to detect
Jun Wang et al.
Cell, 183(7), 1867-1883 (2020-11-29)
Biliary atresia (BA) is a severe cholangiopathy that leads to liver failure in infants, but its pathogenesis remains to be fully characterized. By single-cell RNA profiling, we observed macrophage hypo-inflammation, Kupffer cell scavenger function defects, cytotoxic T cell expansion, and deficiency

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service