Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42663

Sigma-Aldrich

Xanthan from Xanthomonas campestris

4,500,000

Synonym(s):

Xanthan gum from Xanthomonas campestris, Gum xanthan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201

form

powder

InChI

1S/C13H10O/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)14-12/h1-8H,9H2

InChI key

GJCOSYZMQJWQCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

As xanthan is a polysaccharide used in many applications such as a food additive, enzyme substrate or rheology modifier, it is useful to have a xanthan standard with a clearly defined narrow molecular weight distribution. Xanthan is produced by fermentation from Xanthomonas campestris.

Application

Xanthan gum is an anionic polysaccharide composed of a β-(1→4)-D-glucopyranose glucan backbone with side chains of (1→3)-α-D-mannopyranose-(2→1)-β-D-glucuronic acid-(4→1)-β-D-mannopyranose on alternating residues. Approximately half of the terminal mannose residues are 4,6-pyruvated while most of the inner mannose residues are 6-acetylated. Its properties make it a useful matrix component for drug delivery systems. It forms stable drug suspensions in aqueous media and soft gels with locust bean gum or guar gum. Xanthan gum mimics the texture of lipids and is used as a control in experiments where signaling pathways initiated by consumption of lipid-containing reagents are investigated in mice.
Xanthan gum is an anionic polysaccharide composed of a ß-(1→4)-D-glucopyranose glucan backbone with side chains of (1→3)-α-D-mannopyranose-(2→1)-ß-D-glucuronic acid-(4→1)-ß-D-mannopyranose on alternating residues. Approximately half of the terminal mannose residues are 4,6-pyruvated while most of the inner mannose residues are 6-acetylated. Its properties make it a useful matrix component for drug delivery systems. It forms stable drug suspensions in aqueous media and soft gels with locust bean gum or guar gum.

Analysis Note

gel-permeation chromatography corresponds

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analyses of polysaccharide fouling mechanisms during crossflow membrane filtration.
Nataraj, S., et al.
Journal of Membrane Science, 308, 152-161 (2008)
Review of xanthan gum production from unmodified starches by Xanthomonas comprestris sp.
Rosalam, S. and England, R.
Enz. Microbiol. Technol., 39, 197-207 (2006)
Tommasina Coviello et al.
Journal of controlled release : official journal of the Controlled Release Society, 119(1), 5-24 (2007-03-27)
Hydrogels are three-dimensional, hydrophilic, polymeric networks, with chemical or physical cross-links, capable of imbibing large amounts of water or biological fluids. Among the numerous macromolecules that can be used for hydrogel formation, polysaccharides are extremely advantageous compared to synthetic polymers
Shazia N Aslam et al.
Current biology : CB, 18(14), 1078-1083 (2008-07-22)
Bacterial pathogens and symbionts must suppress or negate host innate immunity. However, pathogens release conserved oligomeric and polymeric molecules or MAMPs (Microbial Associated Molecular Patterns), which elicit host defenses [1], [2] and [3]. Extracellular polysaccharides (EPSs) are key virulence factors
Maximina H Yun et al.
Plant physiology, 141(1), 178-187 (2006-03-15)
Xanthan is the major exopolysaccharide secreted by Xanthomonas spp. Despite its diverse roles in bacterial pathogenesis of plants, little is known about the real implication of this molecule in Xanthomonas pathogenesis. In this study we show that in contrast to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service