Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41989

Sigma-Aldrich

DD-2,6-Diaminopimelic acid

≥95.0% (TLC)

Synonym(s):

(2R,6R)-2,6-Diaminoheptanedioic acid, D,D-α,ε-Diaminopimelic acid, D-DAP, NSC 206278

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O4
CAS Number:
Molecular Weight:
190.20
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

assay

≥95.0% (TLC)

optical purity

enantiomeric excess: ≥98.0%

SMILES string

OC([C@H](N)CCC[C@@H](N)C(O)=O)=O

InChI

1S/C7H14N2O4/c8-4(6(10)11)2-1-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13)/t4-,5-/m1/s1

InChI key

GMKMEZVLHJARHF-RFZPGFLSSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The stereoisomers of alpha epsilon-diaminopimelic acid. II. Their distribution in the bacterial order Actinomycetales and in certain Eubacteriales.
D S HOARE et al.
The Biochemical journal, 65(3), 441-447 (1957-03-01)
L K Lam et al.
The Journal of biological chemistry, 263(24), 11814-11819 (1988-08-25)
Analogs 1-8 of diaminopimelic acid (DAP) were synthesized and tested for inhibition of purified meso-DAP D-dehydrogenase from Bacillus sphaericus and of LL-DAP epimerase from Escherichia coli. The dehydrogenase was assayed by monitoring NADPH formation spectrophotometrically at 340 nm. N-Hydroxy DAP
The stereoisomers of alpha epsilon-diaminopimelic acid: their distribution in nature and behaviour towards certain enzyme preparations.
D S HOARE et al.
The Biochemical journal, 61(4), 562-568 (1955-12-01)
Limin Zhang et al.
International journal of systematic and evolutionary microbiology, 62(Pt 2), 272-278 (2011-03-15)
An actinomycete, strain FXJ6.011(T), was isolated from a cup-shaped sponge collected at Dachan reef, Yangpu in the South China Sea. The strain had morphological characteristics of members of the family Micromonosporaceae. Phylogenetic analysis of the 16S rRNA gene sequence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service