Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

417M-1

Sigma-Aldrich

GATA1 (4F5) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4F5, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (417M-14)
vial of 0.5 mL concentrate (417M-15)
bottle of 1.0 mL predilute (417M-17)
vial of 1.0 mL concentrate (417M-16)
bottle of 7.0 mL predilute (417M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG2b

control

bone marrow

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... GATA1(2623)

General description

GATA-1 is a nuclear transcription factor that belongs to the family of GATA proteins which bind the DNA sequence (A/T)GATA-(A/G) by a highly conserved zinc finger domain located on the X chromosome.1,2 GATA1 is expressed in the cells of the erythroid, megakaryocytic and mast cell lineages and is found in low levels in multipotential and Sertoli cells of the testis.1 GATA1 normally suppresses the proliferation of megakaryocytic and erythroid precursors while promoting their differentiation. GATA-1 is crucial for normal erythroid and megakaryocytic development. Missense mutations in GATA1 in humans causes a familial dyserythropoietic anemia and thrombocytopenia. GATA1 has been implicated in childhood acute megakaryoblastic leukemia (AMKL) and transient myeloproliferative disorder in children with Down syndrome (DS). Acquired mutations in exon 2 of GATA1 an X-linked gene has been described in DS patients with AMKL.2 Anti-GATA1 can be used to identify megakaryocytes, erythroid precursors and megakaryocytic malignancies.1, 2

Quality


IVD

IVD

IVD

RUO

Linkage

GATA1 Positive Control Slides, Product No. 417S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liat Rainis et al.
Blood, 102(3), 981-986 (2003-03-22)
Patients with Down syndrome (DS) frequently develop 2 kinds of clonal megakaryocytosis: a common, congenital, spontaneously resolving, transient myeloproliferative disorder (TMD) and, less commonly, childhood acute megakaryoblastic leukemia (AMKL). Recently, acquired mutations in exon 2 of GATA1, an X-linked gene
A G Elefanty et al.
The EMBO journal, 15(2), 319-333 (1996-01-15)
The nuclear distribution of GATA transcription factors in murine haemopoietic cells was examined by indirect immunofluorescence. Specific bright foci of GATA-1 fluorescence were observed in erythroleukaemia cells and primary murine erythroblasts and megakaryocytes, in addition to diffuse nucleoplasmic localization. These

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service