Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41486

Sigma-Aldrich

L-Histidinol phosphate lithium salt

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N3O4P · xLi+
Molecular Weight:
221.15 (free acid basis)
UNSPSC Code:
12352202

assay

≥95% (HPLC)

storage temp.

−20°C

Biochem/physiol Actions

Central metabolite in the histidine biosynthesis pathway of bacteria, yeasts and plants, substrate for histidinolphosphate amino-transferase[1], structure of covalent complex with histidinol-phosphate aminotransferase[2]. Substrate for L-histidinol phosphate phosphatase in the penultimaze step of L-histidine biosynthesis[3].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Sivaraman et al.
Journal of molecular biology, 311(4), 761-776 (2001-08-24)
The biosynthesis of histidine is a central metabolic process in organisms ranging from bacteria to yeast and plants. The seventh step in the synthesis of histidine within eubacteria is carried out by a pyridoxal-5'-phosphate (PLP)-dependent l-histidinol phosphate aminotransferase (HisC, EC
Swapnil V Ghodge et al.
Biochemistry, 52(6), 1101-1112 (2013-01-19)
L-Histidinol phosphate phosphatase (HPP) catalyzes the hydrolysis of L-histidinol phosphate to L-histidinol and inorganic phosphate, the penultimate step in the biosynthesis of L-histidine. HPP from the polymerase and histidinol phosphatase (PHP) family of proteins possesses a trinuclear active site and
Francisco J Fernandez et al.
The Journal of biological chemistry, 279(20), 21478-21488 (2004-03-10)
In histidine biosynthesis, histidinol-phosphate aminotransferase catalyzes the transfer of the amino group from glutamate to imidazole acetol-phosphate producing 2-oxoglutarate and histidinol phosphate. In some organisms such as the hyperthermophile Thermotoga maritima, specific tyrosine and aromatic amino acid transaminases have not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service